Nessler reagent Folin-Wu Modified for determination of non-protein nitrogen (NPN)
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Nota 1
, Potassium tetraiodomercurate
, Mercury (II) potassium iodide
, Potassium tetraiodomercurate – Potassium hydroxide solution
, Potassium mercuric Iodide
R5260000-500B
RC5260-16EA
159.03
USD
RC5260-16
Nessler reagent Folin-Wu Modified for determination of non-protein nitrogen (NPN)
Nessler's reagent
Folin-Wu modified for determination of non-protein nitrogen.
MW:
786.4 g/mol Boiling Pt: 356.6 °C (1013 hPa) Melting Pt: –38.9 °C Density: 13.6 g/cm³ (20 °C) |
MDL Number:
MFCD00049656 EINECS: 231-990-4 UN: 2922 ADR: 8,II Merck Index: 12,07862 |
Specification Test Results
Ammonia Sensitivity | 0.1 mg/L Ammonia Nitrogen |
Appearance | Clear, light yellow, odorless |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...