Molybdate reagent,Taylor type (1.2% Na₂MoO₄) (w/v) in sulfuric acid (4.9%)

Supplier: Ricca Chemical
Danger

R5190000-4A R5190000-10F R5190000-1A R5190000-20F
RC51901EA 239 USD
RC51901 RC519025 RC519032 RC51905
Molybdate reagent,Taylor type (1.2% Na₂MoO₄) (w/v) in sulfuric acid (4.9%)
Molybdate reagent,Taylor type
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, slight greenish-yellow, odorless
Assay at 20 °C (traceable to NIST SRM 84) 1.75 ± 0.02 N at 20.0 °C (Sulfuric Acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR