Magnesium sulfate, anhydrous, powder, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: MgSO4Magnesium sulfate anhydrousMagnesium sulphate anhydrous

Management of Change (MOC) category = N

MACR6070-12 MACR6070-12 MACR6070-10 MACR6070-10 MACR6070-26 6070-26
MK607010CS 6093.18 USD
MK607010 MK607012 CAMK607010 MK607026 CAMK607026 CAMK607012
Magnesium sulfate, anhydrous, powder, AR®, Macron Fine Chemicals™
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://us.vwr.com/stibo/web/std.lang.all/06/17/16640617.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ammonium (NH₄) ≤0.005 %
Arsenic (As) ≤0.001 %
Calcium (Ca) ≤0.04 %
Chloride (Cl) ≤0.001 %
Heavy Metals (as Pb) ≤0.001 %
Insoluble Matter ≤0.01 %
Trace Impurities - Iron (Fe) ≤0.001 %
Loss on Ignition ≤2.0 %
Thru U.S. No. 200 Sieve ≥80 %
Nitrate (NO₃) ≤0.005 %
pH of 5% Solution 5.0 - 9.2
Appearance (fine, white powder, free from lumps)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR