Ertapenem sodium salt ≥97.0%
Supplier: RPI
Synonyms:
Ertapenem sodium, Sodium (4R,5S,6S)-3-(((3S,5S)-5-((3-carboxyphenyl)carbamoyl)pyrrolidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
E32100-1.0
E32100-25.0
E32100-5.0
76345-876EA
169.34
USD
76345-876
76345-878
76345-880
Ertapenem sodium salt ≥97.0%
Ertapenem sodium salt
- Appearance: White to light yellow powder
Formula:
C₂₂H₂₄N₃NaO₇S MW: 497.498 g/mol |
MDL Number:
MFCD09475566 CAS Number: 153773-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...