2-Propanol ≥99.9%, B&J Brand™ for applications requiring low water content, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Isopropyl alcohol
, Isopropyl Rubbing Alcohol
, sec-Propanol
, Isopropyl rubbing alcohol
, 2-Hydroxy propane
, sec-Propyl Alcohol
, Dimethyl carbinol
, 2-Hydroxypropane
, Propane, 2-hydroxy-
, Propan-2-ol
, IPA
, Isopropanol
324-4
324-4
BJ324-4CS
1710.64
USD
CABJ324-4
BJ324-4
2-Propanol ≥99.9%, B&J Brand™ for applications requiring low water content, Burdick & Jackson™
2-Propanol
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Formula:
(CH₃)₂CHOH MW: 60.1 g/mol Boiling Pt: 82.3 °C Melting Pt: -88 °C Density: 0.785 g/cm³ (20 °C) Flash Pt: 12.0 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II REACH: 01-2119457558-25 Merck Index: 13,05228 |
Specification Test Results
GC Analysis | 99.9 % min. |
Refractive Index (20ºC) | 1.3766 - 1.3778 |
Residue | 1 mg/L max. |
UV Absorbance @ 225 nm | 0.16 AU max. |
UV Absorbance @ 250 nm | 0.02 AU max. |
UV Absorbance @ 300 nm | 0.005 AU max. |
UV Absorbance @ 400 nm | 0.005 AU max. |
UV Cutoff | 205 nm max. |
Water by Karl Fischer Titration | 0.01 % max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...