InstaGard® Powder-Free Synthetic Examination Gloves, Cardinal

Supplier: Cardinal Health
8887DOTP 8888DOTP 8889DOTP
PH8887IBCS 568.74 USD
PH8887IB PH8888IB PH8889IB
InstaGard® Powder-Free Synthetic Examination Gloves, Cardinal
Gloves
Ambidextrous gloves offer excellent tactile sensitivity and maximum dexterity.

  • Powder-free
  • Beaded cuff
  • Bisque finish

Seamless vinyl construction provides superior tear resistance.

Color: natural white.

Certifications: Gloves exceed ASTM D5250.

Ordering information: All sizes are packaged in dispenser packs of 100 gloves, except X-large, which is packaged in dispenser packs of 90 gloves.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR