(±)-α-Terpineol ≥97%, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol
, Terpineol
, α-Terpineol
TX0075-1
EM-TX0075-1EA
3956.66
USD
EM-TX0075-1
(±)-α-Terpineol ≥97%, Sigma-Aldrich®
(±)-α-Terpineol
Formula:
C₁₀H₁₈O MW: 154.25 g/mol |
MDL Number:
MFCD00001557 CAS Number: 98-55-5 EINECS: 202-680-6 |
Specification Test Results
Assay (GC) | 97% min |
Color (APHA) | 10 max |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...