Hydrofluoric acid 48%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

MACR2640-18 MACR2640-08 2640-12 MACR2640-12 MACR2640-08 MACR2640-18 MACR2640-12
MK264012-UCS 3351.27 USD
MK264012-U MK264008 CAMK264012 MK2640-18 CAMK264018 CAMK264008 MK264012
Hydrofluoric acid 48%, AR® ACS, Macron Fine Chemicals™
Hydrofluoric acid

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: HF
MW: 20.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HF) (by acidimetry) 48.8 - 49.2 %
ACS - Fluosilicic Acid (H₂SiF₆) (in %) ≤0.01 %
ACS - Residue after Ignition ≤5 ppm
Trace Impurities - Aluminum (Al) ≤0.050 ppm
Trace Impurities - Arsenic (As) ≤0.050 ppm
Trace Impurities - Boron (B) ≤0.050 ppm
Trace Impurities - Calcium (Ca) ≤0.300 ppm
Chloride (Cl) ≤5 ppm
Trace Impurities - Chromium (Cr) ≤0.010 ppm
Trace Impurities - Copper (Cu) ≤0.050 ppm
Trace Impurities - Gold (Au) ≤0.300 ppm
Trace Impurities - Heavy Metals ≤0.5 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm
Trace Impurities - Lead (Pb) ≤0.100 ppm
Trace Impurities - Magnesium (Mg) ≤0.20 ppm
Trace Impurities - Manganese (Mn) ≤0.200 ppm
Trace Impurities - Nickel (Ni) ≤0.1 ppm
Phosphate (PO₄) ≤1 ppm
Trace Impurities - Potassium (K) ≤0.300 ppm
Trace Impurities - Sodium (Na) ≤0.300 ppm
Sulfate and Sulfite (as SO₄) ≤5 ppm
Trace Impurities - Tin (Sn) ≤0.300 ppm
Trace Impurities - Titanium (Ti) ≤0.300 ppm
Trace Impurities - Zinc (Zn) ≤0.300 ppm
Appearance (colorless, corrosive, fuming solution)
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR