Hexane (mixture of isomers) ≥95% (by GC, sum of isomers + methylcyclopentane), J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Hexanes
, Isohexane
Management of Change (MOC) category = N
N169-08
BAKRN169-08
JTN169-8CS
610.85
USD
JTN169-8
CAJTN169-8
Hexane (mixture of isomers) ≥95% (by GC, sum of isomers + methylcyclopentane), J.T.Baker®
Hexane (mixture of isomers)
Formula:
H₃C(CH₂)₄CH₃ MW: 86.18 g/mol Boiling Pt: 68…70 °C (1013 hPa) Melting Pt: –95 °C Density: 0.66 g/cm³ (20 °C) Flash Pt: –22 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179311 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (sum of 5 isomers, total hexanes, plus methylcyclopentane)(by GC) | ≥95 % |
Identification (by IR) | Passes Test |
Appearance (Clear, colorless liquid) | Passes Test |
Color (APHA) | ≤10 |
Residue after Evaporation | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...