STANSOLV® Unlined Nitrile Gloves, Mapa Professionnel
Supplier: Mapa Professionnel
STANSOLV®
470421 479411 480438ZQK 470420 480437ZQK 474017 470427 480430ZQK 470428 479416 479410ZQK 474011 474010 479418ZQK 479419ZQK 479417ZQK 470429
32929-820CS 270.67 USD
32929-820 32929-863 32929-835 32929-834 32928-080 32929-580 32917-853 32917-842 32917-820 32917-831 32917-810 32928-168 32928-146 32928-102 32928-124 32929-808 32928-088
STANSOLV® Unlined Nitrile Gloves, Mapa Professionnel
Gloves
Hand and product protection for food processing, meat packing, agricultural chemical application, electronic assembly and packaging, and acid etching.
StanSolv® gloves have a case-hardened finish to withstand solvents, oil, grease, fats, acids, alkalis, and alcohols. Nitrile (NBR) formulation free of inert fillers and silicone for superior chemical resistance. Gloves contain no natural rubber proteins and will not cause allergic reactions. Also resist punctures, snags, cuts, and abrasion. Hand-specific gloves feature curved finger, tapered wrist, and flat cuff for natural, comfortable wear. Easy donning and doffing. Embossed Z-pattern grip, powder-free.
Certifications: In compliance with 21 CFR 170-199.
Caution: Note: Not recommended for use with ketones, strong oxidizing acids, and organic chemicals containing nitrogen.