Florisil® 60-100 mesh, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: TalcFlorisilSilicic acid magnesium saltMagnesium silicate, activated

Management of Change (MOC) category = N

MACR6875-04 MACR6875-04
MK687504CS 2043.78 USD
MK687504 CAMK6875-04
Florisil® 60-100 mesh, Macron Fine Chemicals™
Magnesium silicate
Formula: MgO₃Si
MW: 100.38 g/mol
Density: 2.51 g/cm³
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
On U.S. No. 60 Sieve ≤10 %
Thru U.S. No. 100 Sieve ≤10 %
Appearance (fine, white crystalline powder). However, due to the high temperature involved in the activation process, some small
charred particles may be visible within the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR