Formaldehyde 37% in aqueous solution stabilized ACS

Supplier: Epredia
Danger


Stabilization: Stabilized with methanol

9300-5 9300-1
48218-722EA 201.86 USD
48218-722 48218-724
Formaldehyde 37% in aqueous solution stabilized ACS
Formaldehyde

Not intended for use as a disinfectant.

Formula: CH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 2209
ADR: 8,III

Order Now

Specification Test Results

Assay, Formaldehyde (wt.) 36.5% - 38.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR