Ethylenediamine ≥98%, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: 1,2-Diaminoethane1,2-Ethanediamine

EX0510-1 EX0510-1
EM-EX0510-1EA 491.31 USD
CAEX0510-1 EM-EX0510-1
Ethylenediamine ≥98%, Supelco®
Ethylenediamine
Formula: C₂H₈N₂
MW: 60.1 g/mol
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: 8.5 °C
Density: 0.898 g/cm³ (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8,II
Merck Index: 13,03829
https://us.vwr.com/stibo/web/std.lang.all/99/69/2299969.jpg

Order Now

Specification Test Results

Assay 98% min
Form Clear liquid, may form carbonates

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR