N-Ethyldiisopropylamine ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
N,N-Diisopropylethylamine
CAS Number: 7087-68-5
MDL Number: MFCD00008868
Molecular Formula: C8H19N
Molecular Weight: 129.25
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 127
Melting point (°C): -45
Flash Point (°C): 12
Specific Gravity (20/20): 0.76
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: N-Ethyldiisopropylamine, DIPEA, Hunig's Base, DIEA
Formula:
[(CH₃)₂CH]₂NC₂H₅ MW: 129.25 g/mol Boiling Pt: 127 °C Melting Pt: –45 °C Density: 0.76 (20 °C) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008868 CAS Number: 7087-68-5 EINECS: 230-392-0 UN: 2733 ADR: 3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...