N,N-Dimethylacetamide ≥99.0%

Supplier: TCI America
Danger

Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

D0641-25ML D0641-500ML
TCD0641-025MLEA 24 USD
TCD0641-025ML TCD0641-500ML
N,N-Dimethylacetamide ≥99.0%
N,N-Dimethylacetamide

CAS Number: 127-19-5 MDL Number: MFCD00008686 Molecular Formula: C4H9NO Molecular Weight: 87.12 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 165 Melting point (°C): -20 Flash Point (°C): 66 Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: N-Acetyldimethylamine, DMA

Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 165 °C
Melting Pt: –20 °C
Density: 0.94 (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d42c87d67b1d72dea40510f6fb4e26fc85d45278&vid=d42c87d67b1d72dea40510f6fb4e26fc85d45278&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR