BioX Tissue Culture Flasks
Supplier: TWD Scientific
BX07-8025 BX07-8075 BX07-8182 BX07-8300
77394-954CS 215.95 USD
77394-954 77394-956 77394-958 77394-960
BioX Tissue Culture Flasks
Flasks Culture Flasks
Ideal for the cultivation and observation of sensitive cells.
  • Tilted neck
  • PTFE hydrophobic vent cap
  • Marked volume scales on both sides
  • Frosted writing area
  • DNase/RNase, non-pyrogenic
  • Highly transparent polystyrene
The angled neck allows the user to access the entire surface area. Easy sample identification due to the molded graduation and frosted writing patch.