Hot Start High Fidelity DNA Polymerase 2X master Mix
Supplier: G-Biosciences
786-1972 786-1973
76716-654EA 174.64 USD
76716-654 76716-696
Hot Start High Fidelity DNA Polymerase 2X master Mix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Hot start high-fidelity DNA Polymerase 2X master mix is ready to use premix which contains high fidelity DNA Polymerase, dNTPs and optimized buffer with stabilizers. The high-fidelity DNA Polymerase possesses 5´→3´ polymerase and 3´→5´ exonuclease (proofreading) activities which has the fidelity, sensitivity and processivity with an error rate ~2.8×10² fold lower than Taq DNA polymerase, and significantly lower than other proofreading DNA polymerases.
  • High fidelity PCR and cloning
Long and high GC content template DNA amplification.