High Fidelity DNA Polymerase 2X Master Mix
Supplier: G-Biosciences
786-1968
786-1969
76716-682EA
153.31
USD
76716-682
76716-640
High Fidelity DNA Polymerase 2X Master Mix
Nucleic Acid Reagents
End-point PCR Enzymes and Kits
High-fidelity DNA Polymerase 2x master mix is ready to use premix which contains high fidelity DNA Polymerase, dNTPs and optimized buffer with stabilizers. The high-fidelity DNA Polymerase possesses 5´→3´ polymerase and 3´→5´ exonuclease (proofreading) activities which has the fidelity, sensitivity and processivity with an error rate ~2.8×10² fold lower than Taq DNA polymerase, and significantly lower than other proofreading DNA polymerases.
- High fidelity PCR and cloning
Long and high GC content template DNA amplification.