Celite® 545, filter aid, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: DE, Diatomaceous earth, flux-calcined

CX0574-3 CX0574-7 CX0574-1
EM-CX0574-3EA 115 USD
EM-CX0574-3 EM-CX0574-7 EM-CX0574-1
Celite® 545, filter aid, Supelco®
Celite® 545, filter aid
Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00132803
CAS Number: 68855-54-9
EINECS: 272-489-0

Order Now

Specification Test Results

Color Off-white
Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR