Dichloroacetic acid (DCA) 3% in toluene, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: DCADCA Deblock

SR674-4 SR674-4
BJSR674-4CS 971.68 USD
BJSR674-4 CABASR6744TWT
Dichloroacetic acid (DCA) 3% in toluene, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Dichloroacetic acid (DCA)

Reagents are specifically processed and purified to ensure the high synthesis efficiency


  • Ideal for oligonucleotide synthesis
  • Extremely low water content
  • High purity reagents
  • Low amine specifications


Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.

Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Density: 8,815 g/cm3
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR