Premium Natural Rubber Latex Gloves, Unsupported, Honeywell Safety
Supplier: Honeywell Safety
PNLB1815/O/10 PNLB1815/O/10 PNLB1815/0/8 PNLB1815/O/9 PNLB1815/O/8
32888-303CS 1132.32 USD
CA32888-303 CA32888-304 32888-303 CA32888-306 32888-306
Premium Natural Rubber Latex Gloves, Unsupported, Honeywell Safety
Gloves
High-quality, natural rubber latex gloves offer excellent resistance against abrasions, snags, and cuts.
  • Case hardened for easy donning/doffing
  • Positive grip in wet/dry applications
  • Beaded cuff for increased strength and tear resistance
  • Size embossed instead of inked to eliminate ink contamination
  • Chlorinated for more comfort and ease of use
  • Powder-free, color: orange
Gloves are excellent for handling tasks and protection from acids, caustics, animal fats, and vegetable oils. Used in food handling, pharmaceutical, and electronics applications.
Certifications: Meet ASTM 739 permeation standard. Comply with USDA and FDA regulations CFR 21 for use in food processing.