Cesium chloride 99.9%, powder, Optical Grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Cesium chloride is used for the preparation of electrically conducting glasses.
Used for density gradient work and for the purification of virus/phage, nucleic acids and nucleoproteins
Cesium chloride is used for the preparation of electrically conducting glasses. Used to make solutions for the separation of RNA and DNA, virus/phage by density gradient centrifugation.
Freely soluble in water (50% aqueous - clear, colorless solution); soluble in ethanol, alcohol.
Formula:
CsCl Boiling Pt: 1303 °C (1013 hPa) Melting Pt: 645 °C (lit.) Density: 3.7808 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010955 CAS Number: 7647-17-8 EINECS: 231-600-2 Merck Index: 13,02021 |
Specification Test Results
Purity | 99.9% |
pH (1% aq soln) | 5.5 - 6.5 |
Barium | ≤10 ppm |
Magnesium | ≤10 ppm |
Potassium | ≤30 ppm |
Iron | ≤10 ppm |
Calcium | ≤10 ppm |
Solubility (50% aq soln) | Clear and Colorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...