VWR® Glass Pennyhead Stoppers, [ST]
Supplier: Avantor
VWR®
012.205.07 012.205.04 012.205.00
77441-922CS 840 USD
77441-922 77441-920 77441-918
VWR® Glass Pennyhead Stoppers, [ST]
Stoppers Glass Stoppers
Solid 3.3 borosilicate glass stoppers.
  • Pennyhead style
For use with round bottom flasks, Erlenmeyer flasks, and separating funnels.