1,4-Butanediolbis(3-aminopropyl)ether ≥98.0%
Supplier: TCI America
Synonyms:
1,4-Bis(3-aminopropoxy)butane
CAS Number: 7300-34-7
MDL Number: MFCD00009802
Molecular Formula: C10H24N2O2
Molecular Weight: 204.31
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 159
Specific Gravity (20/20): 0.96
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Bis(3-aminopropoxy)butane, 4,9-Dioxa-1,12-dodecanediamine
Formula:
C₁₀H₂₄N₂O₂ MW: 204.31 g/mol Boiling Pt: 135 °C Density: 0.96 (20 °C) Flash Pt: 159 °C |
MDL Number:
MFCD00009802 CAS Number: 7300-34-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...