Benzyl alcohol ≥99.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: PhenylmethanolBenzenemethanolPhenylcarbinol

BX0483-3 BX0483-3
EM-BX0483-3CS 6077 USD
CABX0483-3 EM-BX0483-3
Benzyl alcohol ≥99.0%, GR ACS, Supelco®
Benzyl alcohol

Meets ACS Specifications

Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15.3 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://us.vwr.com/stibo/web/std.lang.all/97/87/2299787.jpg

Order Now

Specification Test Results

Assay (GC) 99.0% min
Acetophenone 0.02% max
Acidity 0.01meq/g max
Benzaldehyde 0.01% max
Chlorinated compounds To pass test
Color (APHA) 20 max
Form Clear liquid
Identification To pass test
Refractive index (n 20/D) 1.539-1.541
Residue after evaporation 0.05% max
Residue after ignition 0.005% max
Specific Gravity at 25/25°C 1.042-1.047

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR