6-Fluoro-2-methyl-3-nitrobenzyl alcohol 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: (6-Fluoro-2-methyl-3-nitrophenyl)methanol

PC56307-1G PC56307-5G
77431-874EA 58.08 USD
77431-874 77431-876
6-Fluoro-2-methyl-3-nitrobenzyl alcohol 97%
6-Fluoro-2-methyl-3-nitrobenzyl alcohol
Formula: C₈H₈FNO₃
Storage Temperature: Ambient
MDL Number: MFCD28347725
CAS Number: 1613329-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR