3'-Azido-3'-deoxythymidine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Azidothymidine1-(3-Azido-2,3-dideoxy-beta-D-ribofuranosyl)thymineAZTZidovudine

OR8250T-250MG
77424-474EA 13 USD
77424-474
3'-Azido-3'-deoxythymidine 97%
3'-Azido-3'-deoxythymidine

A potent & selective inhibitor of HIV-1 replication.

Formula: C₁₀H₁₃N₅O₄
MW: 267.25 g/mol
Melting Pt: 113…115 °C
Storage Temperature: Freezer
MDL Number: MFCD00006536
CAS Number: 30516-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR