3-Isopropylaniline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Aminocumenem-Cumidine

OR0560-1G
77402-128EA 16 USD
77402-128
3-Isopropylaniline
3-Isopropylaniline
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 101…103 °C/15 mmHg
Density: 0.96 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00092918
CAS Number: 5369-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR