Chlorobenzene
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Phenylchloride, Benzene Chloride, Monochlorobenzene
OR8791-100ML
OR8791-250ML
77411-442EA
21.32
USD
77411-442
77411-444
Chlorobenzene
Chlorobenzene
Formula:
C₆H₅Cl Boiling Pt: 132 °C Melting Pt: –45 °C Density: 1.106 g/cm³ (25 °C) Flash Pt: 27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000530 CAS Number: 108-90-7 EINECS: 203-628-5 UN: 1134 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...