2-Fluoro-5-(trifluoromethyl)benzyl alcohol 80%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC99525-100G
PC99525-25G
PC99525-5G
77418-848EA
239.92
USD
77418-848
77418-850
77418-852
2-Fluoro-5-(trifluoromethyl)benzyl alcohol 80%
2-Fluoro-5-(trifluoromethyl)benzyl alcohol
Formula:
C₈H₆F₄O |
MDL Number:
MFCD00061167 CAS Number: 207974-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...