Methyl 2,6-difluoro-3-(trifluoromethoxy)benzoate 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC57428-1G PC57428-250MG PC57428-5G
77432-100EA 496.16 USD
77432-100 77432-102 77432-104
Methyl 2,6-difluoro-3-(trifluoromethoxy)benzoate 95%
Methyl 2,6-difluoro-3-(trifluoromethoxy)benzoate
Formula: C₉H₅F₅O₃
MDL Number: MFCD34181141
CAS Number: 2676196-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR