3-(1,1-Difluoroethyl)benzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC52530-1G PC52530-250MG PC52530-5G
77431-308EA 330.1 USD
77431-308 77431-310 77431-312
3-(1,1-Difluoroethyl)benzonitrile 95%
3-(1,1-Difluoroethyl)benzonitrile
Formula: C₉H₇F₂N
MDL Number: MFCD27925456
CAS Number: 55805-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR