1-Chloro-5-fluoro-4-methoxy-2-nitrobenzene

Supplier: APOLLO SCIENTIFIC
Warning

PC104079-1G PC104079-5G
77426-618EA 69.28 USD
77426-618 77426-620
1-Chloro-5-fluoro-4-methoxy-2-nitrobenzene
1-Chloro-5-fluoro-4-methoxy-2-nitrobenzene
Formula: C₇H₅ClFNO₃
MDL Number: MFCD29043481
CAS Number: 98404-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR