3-bromo-2-hydroxy-5-iodobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Brom-5-iod-salicylsaeure

OR97135-1G OR97135-25G OR97135-5G
77425-950EA 496.16 USD
77425-950 77425-952 77425-954
3-bromo-2-hydroxy-5-iodobenzoic acid
3-bromo-2-hydroxy-5-iodobenzoic acid
Formula: C₇H₄BrIO₃
MDL Number: MFCD26396222
CAS Number: 4068-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR