2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,2'-Azobis[2-(2-imidazolin-2-yl)propane] Dihydrochloride, 1,2-Bis(2-(4,5-dihydro-1H-imidazol-2-yl)propan-2-yl)diazene dihydrochloride, 2-{2-[(E)-2-[2-(4,5-Dihydro-1H-imidazol-2-yl)proan-2-yl]diazen-1-yl]propan-2-yl}-4,5-dihydro-1H-imidazole dihydrochloride

OR917769-100G OR917769-25G
77424-952EA 166.06 USD
77424-952 77424-954
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride 98%
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride
Formula: C₁₂H₂₄Cl₂N₆
Melting Pt: 190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142723
CAS Number: 27776-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR