2-Bromo-3-iodopyridin-4-amine 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR62195-1G OR62195-250MG
77423-706EA 329.08 USD
77423-706 77423-708
2-Bromo-3-iodopyridin-4-amine 95%
2-Bromo-3-iodopyridin-4-amine
Formula: C₅H₄BrIN₂
MDL Number: MFCD28344964
CAS Number: 1300750-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR