2-Aminoethyl methacrylate hydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Aminoethyl 2-methylprop-2-enoate hydrochloride

OR61529-10G OR61529-1G OR61529-5G
77423-604EA 74.98 USD
77423-604 77423-606 77423-608
2-Aminoethyl methacrylate hydrochloride 98%
2-Aminoethyl methacrylate hydrochloride
Formula: C₆H₁₂ClNO₂
MW: 165.617 g/mol
Melting Pt: 102…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078260
CAS Number: 2420-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR