4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4'-[(6-Acryloyloxy)hexyloxy]-4-biphenylcarbonitrile, 6-[(4'-Cyanobiphenyl-4-yl)oxy]hexyl acrylate, Acrylic acid 6-[(4'-Cyano-[1,1'-biphenyl]-4-yl)oxy]hexyl ester, 6-[(4'-Cyano-[1,1'-biphenyl]-4-yl)oxy]hexyl acrylate, 6-((4'-Cyano-[1,1'-biphenyl]-4-yl)oxy)hexyl acrylate

OR53136-1G OR53136-25G OR53136-5G
77422-806EA 48.9 USD
77422-806 77422-808 77422-810
4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl 98%
4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl
Formula: C₂₂H₂₃NO₃
Melting Pt: 71 °C
MDL Number: MFCD16293619
CAS Number: 89823-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR