Pyridine-2-carbaldehyde 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR5051-100G
OR5051-25G
OR5051-500G
77410-114EA
117.16
USD
77410-114
77410-116
77410-118
Pyridine-2-carbaldehyde 99%
Pyridine-2-carbaldehyde
Formula:
C₆H₅NO Boiling Pt: 179…181 °C Melting Pt: –21 °C Density: 1.126 Flash Pt: 73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006290 CAS Number: 1121-60-4 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...