4-Acetylphenylboronic acid pinacol ester 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone, 4-Acetylphenylboronic acid, pinacol ester, 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone

OR360618-1G OR360618-25G OR360618-5G
77420-614EA 24.46 USD
77420-614 77420-616 77420-618
4-Acetylphenylboronic acid pinacol ester 95%
4-Acetylphenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₃
Boiling Pt: 356.6/760mm °C
Melting Pt: 63…65 °C
Density: 1.04
Flash Pt: 169.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05863922
CAS Number: 171364-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR