Tetraphenylphosphonium bromide 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR315491-100G OR315491-25G OR315491-500G
77409-082EA 55.02 USD
77409-082 77409-084 77409-086
Tetraphenylphosphonium bromide 98%
Tetraphenylphosphonium bromide
Formula: C₂₄H₂₀BrP
Melting Pt: 294…296 °C
MDL Number: MFCD00011915
CAS Number: 2751-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR