3'-Aminoacetanilide 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-(3-Aminophenyl)acetamide, 3-Acetamidoaniline, N-Acetyl-1,3-phenylenediamine, m-Aminoacetanilide

OR30419-100G OR30419-25G
77413-526EA 23.4 USD
77413-526 77413-528
3'-Aminoacetanilide 98%
3'-Aminoacetanilide
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Boiling Pt: 388
Melting Pt: 84…88 °C
MDL Number: MFCD00025229
CAS Number: 102-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR