Transport Tote XL Tube Rack Inserts
Supplier: Heathrow Scientific
120960 120961 120962
77395-618CS 97.91 USD
77395-618 77395-620 77395-622
Transport Tote XL Tube Rack Inserts
Racks Tube Racks
Inserts designed to fit the Transport tote XL sample transport container.
  • Thermoformed PETG construction
  • Tube Rack Inserts can be used separately
  • Lid removes completely for easy cleaning
  • Inserts are deep enough to be filled with ice
  • Non sterile
Rack inserts are available in small tube, large tube, and multi-tube formats, accommodating a wide variety of tube sizes and placement configurations.