L(-)-Glutathione (reduced form)
Supplier: CUBE BIOTECH
Synonyms:
L-Glutathione Reduced
, Glutathione Reduced
, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid
, GSH
Reduced L-Glutathione ligand is, therefore, an essential component for GST-tag-based protein purification assays. Loose L-Glutathione (reduced) is a compound of the elution buffer of GST-tag purification assays. It works by giving the GST-tag an alternative substrate to bind to instead of the L-Glutathione on the involved purification beads.
Appearance: White powder
Amount per Bottle: 25 g
Usage: Competetive elution of GST tagged proteins from GST affinity beads
Assay (HPLC): Min. 97%
Spec. rotation ([α]20 D): -16° - -19° (2% in water)
Soluble in 5% in water: Clear, colourless solution
Solubility: Approx. 100 g/l (H2O)
Heavy Metals (as Pb): Max. 0.0005%
Formula: C10H17N3O6S
Formula:
C₁₀H₁₇N₃O₆S MW: 307.33 g/mol |
MDL Number:
MFCD00065939 CAS Number: 70-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...