4-(Trifluoromethylthio)phenol 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-[(Trifluoromethyl)sulphanyl]phenol4-[(Trifluoromethyl)sulfanyl]phenol

PC7773A-100G PC7773A-25G PC7773A-5G
77433-370EA 16.48 USD
77433-370 77433-372 77433-368
4-(Trifluoromethylthio)phenol 97%
4-(Trifluoromethylthio)phenol
Formula: C₇H₅F₃OS
MW: 194.18 g/mol
Boiling Pt: 77…78 °C
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00036035
CAS Number: 461-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR