3-Amino-2-fluoropyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Fluoro-3-pyridylamine, 2-Fluoropyridin-3-amine

PC9189-1G PC9189-5G
77434-240EA 21.28 USD
77434-240 77434-242
3-Amino-2-fluoropyridine
3-Amino-2-fluoropyridine
Formula: C₅H₅FN₂
MW: 112.11
Density: 1.212 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03095248
CAS Number: 1597-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR