Perfluorododecane 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC5992D-25G PC5992D-5G
77432-440EA 353.52 USD
77432-440 77432-442
Perfluorododecane 97%
Perfluorododecane
Formula: C₁₂F₂₆
Boiling Pt: 178 °C
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00042084
CAS Number: 307-59-5
EINECS: 206-204-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR