Acetamidoxime

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Acetamide oxime

OR30585-1G OR30585-5G
77418-966EA 20.26 USD
77418-966 77418-968
Acetamidoxime
Acetamidoxime
Formula: C₂H₆N₂O
MW: 74.08 g/mol
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00603514
CAS Number: 22059-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR