Glyoxylic acid monohydrate 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Formylformic acid monohydrateOxoacetic acid monohydrate2-Oxoethanoic acid monohydrate2,2-Dihydroxyacetic acid monohydrate

OR52099-25G OR52099-100G OR52099-500G
77410-206EA 22.66 USD
77410-206 77410-204 77410-208
Glyoxylic acid monohydrate 97%
Glyoxylic acid monohydrate
Formula: C₂H₂O₃·1H₂O
MW: 92.05 g/mol
Boiling Pt: 100 °C
Melting Pt: 49…52 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00127974
CAS Number: 563-96-2
EINECS: 206-058-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR