4-Bromo-2-chloro-3-fluorobenzoic acid 95%

Supplier: APOLLO SCIENTIFIC

PC52713-1G PC52713-250MG PC52713-5G
77431-328EA 500.24 USD
77431-328 77431-330 77431-332
4-Bromo-2-chloro-3-fluorobenzoic acid 95%
4-Bromo-2-chloro-3-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28733905
CAS Number: 1807036-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR